Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Rabbit IgG F(c) Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: ImmunoReagents
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Conjugation: AP (Alkaline Phosphatase)
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1,0 mg 1 mg IMMRIR2130
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol IgG F(c)
Antigen name Immunoglobulin G
Reactivity Rabbit
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antibody type Secondary
Host Goat
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
Epitope F(c)
Concentration 1,50 mg/ml (E 1% at 280 nm = 13,0)
Format Purified IgG
Cross adsorption Yes
Immunogen Rabbit IgG Fc
Purification Affinity purified using solid phaseRabbit IgG
Form Clear, colorless liquid
Antibody fragment F(c)
Storage buffer 30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
Storage temperature 2-8 °C
Shipping temperature 2-8 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty