Digital thermo-hygrometer
Supplier: Amarell
AMARE915200EA 229 EUR
AMARE915200
Digital thermo-hygrometer
Hygrometers Digital Thermo-Hygrometers
This handheld instument is designed to measure relative air humidity and ambient temperature.
  • Precise and fast response
  • Capacitive sensor for humidity
  • Sinter-filter helps measuring in bulk material
W×D×H: 115×65×26 mm