Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calciumglycerophosphate hydrate, EMPROVE® exp Ph. Eur., FCC suitable for use as excipient

Supplier: MERCK PRODUCTION CHEMICALS



Formula: C₃H₇CaO₆P·xH₂O
MDL Number: MFCD00151656
CAS Number: 28917-82-0

 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
25 kg 1.04112.9025
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty