You Searched For: Degassers
Degassers of all models and formulations are designed to effectively remove dissolved or adsorbed gases from HPLC mobile phases. Removing gases is important in eliminating noise spikes in chromatograms. Bubble formation on the mixing of solvents can lead to a number of problems in HPLC analysis which can be prevented by degassing the mobile phase. Degassers may use the processes of helium purging, vacuum degassing, or sonication to achieve positive results.
Degassers of all models and formulations are designed to effectively remove dissolved or adsorbed gases from HPLC mobile phases. Removing gases is important in eliminating noise spikes in chromatograms. Bubble formation on the mixing of solvents can lead to a number of problems in HPLC analysis which can be prevented by degassing the mobile phase. Degassers may use the processes of helium purging, vacuum degassing, or sonication to achieve positive results.
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (RHEO9000-1549)
Supplier: IDEX HEALTH&SCIENCE LLC(RHEODY
Description: Dissolved gases and bubbles in system liquids cause dispense volume anomalies in many instruments, negatively affecting both dispense precision and an...
UOM: 1 * 1 items
Catalog Number: (HPLC0001-6484)
Supplier: COLLECTION PREFIX HPLC
Description: The Systec® Stand-Alone Prep-Scale HPLC vacuum degassing systems is high-efficiency, in-line modules that remove dissolved gases from the mobile phase...
UOM: 1 * 1 items
Supplier: COLLECTION PREFIX HPLC
Description: On-Line degassing has been shown to be more effective at removing dissolved gas from mobile phases than sonication or helium sparging. Up to five solv...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3 of 3
no targeter for Bottom