You Searched For: Selection Valves
Multiple columns in the selection valve allow you to apply different analytical methods while using the same instrument. The common port of this valve connects to the injector. Turning the valve handle directs the flow into one of the columns connected to the peripheral ports. A second valve is placed at the column outlets to select the operating column effluent and direct it to the detector. The valve has a spring-loaded detent mechanism that allows for continuous rotation in either direction.
Multiple columns in the selection valve allow you to apply different analytical methods while using the same instrument. The common port of this valve connects to the injector. Turning the valve handle directs the flow into one of the columns connected to the peripheral ports. A second valve is placed at the column outlets to select the operating column effluent and direct it to the detector. The valve has a spring-loaded detent mechanism that allows for continuous rotation in either direction.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: IDEX HEALTH&SCIENCE LLC(RHEODY
Description: Combine MX Series IIâ„¢ modular valves with an instrument currently in use to support complex fluid switching and sample injection needs.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.