Anti-IgG (H+L) Goat Polyclonal Antibody (Agarose)

Supplier: Rockland Immunochemicals
ROCK210-1150EA 278 EUR
ROCK210-1150
Anti-IgG (H+L) Goat Polyclonal Antibody (Agarose)
Antibodies
Recommended Dilutions:
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR