Anti-sPLA2 Type IIA Mouse Monoclonal Antibody [clone: SCACC353]

Supplier: Cayman Chemical
CAYM160500-1EA 415 EUR
CAYM160500-1
Anti-sPLA2 Type IIA Mouse Monoclonal Antibody [clone: SCACC353]
Antibodies
Type: Primary
Antigen: sPLA2 Type IIA
Clonality: Monoclonal
Clone: SCACC353
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR