Absorbent pads
Supplier: Sartorius
SART15410-047-ALREA 247 EUR
SART15410-047-ALR SART15410-50-ALR
Absorbent pads
Sartorius 1,4 mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come pre-sterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
  • The absorbent pads are available in two diameters:
  • 47 mm with approx. 3 ml absorption capacity
  • 50 mm with approx. 3,5 ml absorption capacity