Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RQCD1 Mouse Monoclonal Antibody

Supplier: Abnova
Mouse monoclonal antibody raised against a full-length recombinant RQCD1.

Type: Primary
Antigen: RQCD1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
Up to 5 Clones 1 ABNOH00009125-M
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol RQCD1
Antigen name RCD1 required for cell differentiation1 homolog (S. pombe)
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Host Mouse
Western blot Yes
ELISA Yes
Antigen synonyms RCD-1, CT129, CNOT9, CAF40, RCD1
Amino acid number 1 to 299
Gene ID 9125
Format Liquid
Cross adsorption No
Immunogen RQCD1 (NP_005435.1, 1 a.a. ~ 299 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Sequence MHSLATAAPVPTTLAQVDREKIYQWINELSSPETRENALLELSKKRESVPDLAPMLWHSFGTIAALLQEIVNIYPSINPPTLTAHQSNRVCNALALLQCVASHPETRSAFLAAHIPLFLYPFLHTVSKTRPFEYLRLTSLGVIGALVKTDEQEVINFLLTTEIIPLCLRIMESGSELSKTVATFILQKILLDDTGLAYICQTYERFSHVAMILGKMVLQLSKEPSARLLKHVVRCYLRLSDNPRAREALRQCLPDQLKDTTFAQVLKDDTTTKRWLAQLVKNLQEGQVTDPRGIPLPPQ
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty