Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IQCB1 Mouse Monoclonal Antibody

Supplier: Abnova
Mouse monoclonal antibody raised against a full-length recombinant IQCB1.

Type: Primary
Antigen: IQCB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
Up to 5 Clones 1 ABNOH00009657-M
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IQCB1
Antigen name IQ motif containing B1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Host Mouse
Western blot Yes
ELISA Yes
Antigen synonyms SLSN5, PIQ, NPHP5
Amino acid number 1 to 465
Gene ID 9657
Format Liquid
Cross adsorption No
Immunogen IQCB1 (NP_001018865.1, 1 a.a. ~ 465 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Sequence MKPTGTDPRILSIAAEVAKSPEQNVPVILLKLKEIINITPLGSSELKKIKQDIYCYDLIQYCLLVLSQDYSRIQGGWTTISQLTQILSHCCVGLEPGEDAEEFYNELLPSAAENFLVLGRQLQTCFINAAKAEEKDELLHFFQIVTDSLFWLLGGHVELIQNVLQSDHFLHLLQADNVQIGSAVMMMLQNILQINRSKRSKMLLEINRQKEEEDLKLQLQLQRQRAMRLSRELQLSMLEIVHPGQVEKHYREMEEKSALIIQKHWRGYRERKNFHQQRQSLIEYKAAVTLQRAALKFLAKYRKKKKLFAPWRGLQELTDARRVELKKRVDDYVRRHLGSPMSDVVSRELHAQAQERLQHYFMGRALEERAQQHREALIAQISTNVEQLMKAPSLKEAEGKEPELFLSRSRPVAAKAKQAHLTTLKHIQAPWWKKLGEESGDEIDVPKDELSIELENLFIGGTKPP
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty