Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTER Mouse Monoclonal Antibody

Supplier: Abnova
Mouse monoclonal antibody raised against a full-length recombinant PTER.

Type: Primary
Antigen: PTER
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
Up to 5 Clones 1 ABNOH00009317-M
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PTER
Antigen name Phosphotriesterase related
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Host Mouse
Western blot Yes
ELISA Yes
Antigen synonyms RPR-1, HPHRP
Amino acid number 1 to 349
Gene ID 9317
Format Liquid
Cross adsorption No
Immunogen PTER (NP_001001484.1, 1 a.a. ~ 349 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Sequence MSSLSGKVQTVLGLVEPSKLGRTLTHEHLAMTFDCCYCPPPPCQEAISKEPIVMKNLYWIQKNAYSHKENLQLNQETEAIKEELLYFKANGGGALVENTTTGISRDTQTLKRLAEETGVHIISGAGFYVDATHSSETRAMSVEQLTDVLMNEILHGADGTSIKCGIIGEIGCSWPLTESERKVLQATAHAQAQLGCPVIIHPGRSSRAPFQIIRILQEAGADISKTVMSHLDRTILDKKELLEFAQLGCYLEYDLFGTELLHYQLGPDIDMPDDNKRIRRVRLLVEEGCEDRILVAHDIHTKTRLMKYGGHGYSHILTNVVPKMLLRGITENVLDKILIENPKQWLTFK
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty