Anti-ADGRL3 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB25970EA 643 EUR
ABNOPAB25970
Anti-ADGRL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LPHN3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-6 ?g/ml

Type: Primary
Antigen: ADGRL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR