Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADAMTS5 Rabbit Polyclonal Antibody

Supplier: Abnova
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS5.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 11 μg/ml

Type: Primary
Antigen: ADAMTS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Gorilla, Human, Monkey, Mouse, Pig, Rat



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 μg 50 µg ABNOPAB25974
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ADAMTS5
Antigen name ADAM Metallopeptidase with Thrombospondin Type 1 Motif 5
Reactivity Gorilla, Pig, Mouse, Monkey, Bovine, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms ADAMTS11, ADAMTS-11, ADMP-2, ADAMTS-5, ADAM-TS 5, ADAM-TS 11, ADAM-TS5
Gene ID 11096
Format Liquid
Cross adsorption No
Immunogen A synthetic peptide corresponding to 20 amino acids at internal region of human ADAMTS5.
Purification Immunoaffinity chromatography
Storage buffer PBS (0,1% sodium azide)
Storage temperature Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty