Anti-ERBB4 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB26002EA 643 EUR
ABNOPAB26002
Anti-ERBB4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ERBB4.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 16 ?g/ml

Type: Primary
Antigen: ERBB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Dog, Horse, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR