Anti-FPR3 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB26177EA 643 EUR
ABNOPAB26177
Anti-FPR3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FPR3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 8 ?g/ml

Type: Primary
Antigen: FPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human, Monkey, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR