Anti-FZD9 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB26261EA 643 EUR
ABNOPAB26261
Anti-FZD9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-10 ?g/ml

Type: Primary
Antigen: FZD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR