Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DYKDDDDK tag Polyclonal Antibody

Supplier: Abnova
Rabbit polyclonal antibody raised against synthetic peptides of DYKDDDDK tag.

Recommended Dilutions: Western Blot: 0.1-1 μg/ml; ELISA: 0.01-0.1 μg/ml; Immunoprecipitation: 2-5 μg/ml

Type: Primary
Antigen: DYKDDDDK tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 μg 100 µg ABNOPAB12767
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol DYKDDDDK
Antigen name DYKDDDDK tag
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Format Liquid
Cross adsorption No
Immunogen A synthetic peptide corresponding to DYKDDDDK tag.
Storage buffer PBS, pH 7,2 (10% Proclin300)
Storage temperature Store at 4 °C. For long term storage store at – 20 °C or lower. Aliquot to avoid repeated freezing and thawing.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping