Heat resistant gloves, RGT 1685
Supplier: Honeywell Safety Products
DALLRGT1685_UEA 0 EUR
DALLRGT1685_U
Heat resistant gloves, RGT 1685
These knitted cotton terry gloves are ideal for handlings of warm parts in glass industries. They provide protection against contact temperatures until 250 °C.
  • Ribbed at the wrist to ensure a good level of wrist comfort, thus eliminating risks of getting it caught
  • 5-finger glove for a better grip of hot objects as well as added dexterity
  • Breathable terry threads provides the glove with better thermal insulation
  • Cotton provides mechanical resistance and insulation
Certifications: EN 388 (1232), EN 407 (X2XXXX); Cat. II