Anti-ZNF232 Antibody Pair

Supplier: Abnova
H00007775-AP51
ABNOH00007775-AP51EA 835 EUR
ABNOH00007775-AP51
Anti-ZNF232 Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human ZNF232.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-ZNF232 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-ZNF232 (50 µg);

Type:
Antigen: ZNF232
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR