Anti-IL1A + IL1R2 Antibody Pair

Supplier: Abnova
DI0530
ABNODI0530EA 794 EUR
ABNODI0530
Anti-IL1A + IL1R2 Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the IL1A protein, and the other against the IL1R2 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. IL1A Rabbit purified Polyclonal Antibody (20 µg) 2. IL1R2 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: IL1A + IL1R2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR