Anti-RPS6KA5 + EGF Antibody Pair

Supplier: Abnova
DI0542
ABNODI0542EA 794 EUR
ABNODI0542
Anti-RPS6KA5 + EGF Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the RPS6KA5 protein, and the other against the EGF protein for use in Proximity Ligation Assay

Antibody pair set content: 1. RPS6KA5 Rabbit purified Polyclonal Antibody (20 µg) 2. EGF Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: RPS6KA5 + EGF
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR