Anti-AK1 Antibody Pair

Supplier: Abnova
H00000203-AP12
ABNOH00000203-AP12EA 884 EUR
ABNOH00000203-AP12
Anti-AK1 Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human AK1.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AK1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-AK1, IgG1 kappa (20 µg);

Type:
Antigen: AK1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR