Anti-SNCG Polyclonal Antibody Pair

Supplier: Abnova
H00006623-AP21
ABNOH00006623-AP21EA 835 EUR
ABNOH00006623-AP21
Anti-SNCG Polyclonal Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human SNCG.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-SNCG (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-SNCG (20 µg);

Type:
Antigen: SNCG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR