Anti-CLEC2D Antibody Pair

Supplier: Abnova
H00029121-AP51
ABNOH00029121-AP51EA 835 EUR
ABNOH00029121-AP51
Anti-CLEC2D Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human CLEC2D.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CLEC2D (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-CLEC2D (50 µg);

Type:
Antigen: CLEC2D
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR