Anti-SULT2B1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00006820-PW1
ABNOH00006820-PW1EA 835 EUR
ABNOH00006820-PW1
Anti-SULT2B1 Rabbit Polyclonal Antibody
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-SULT2B1 (300 ?l); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-SULT2B1 (50 ?l)

Type:
Antigen: SULT2B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR