Anti-CTH Antibody Pair

Supplier: Abnova
H00001491-AP52
ABNOH00001491-AP52EA 835 EUR
ABNOH00001491-AP52
Anti-CTH Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human CTH.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CTH (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-CTH (50 µg);

Type:
Antigen: CTH
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR