Anti-HDAC3 Mouse Monoclonal Antibody (Biotin)

Supplier: Abnova
H00008841-AP45
ABNOH00008841-AP45EA 835 EUR
ABNOH00008841-AP45
Anti-HDAC3 Mouse Monoclonal Antibody (Biotin)
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human HDAC3.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-HDAC3 (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-HDAC3, IgG1 kappa (50 ?g);

Type:
Antigen: HDAC3
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR