Anti-CD74 Antibody Pair

Supplier: Abnova
H00000972-PW2
ABNOH00000972-PW2EA 884 EUR
ABNOH00000972-PW2
Anti-CD74 Antibody Pair
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-CD74 (300 µl); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-CD74 (50 µl)

Type:
Antigen: CD74
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR