Anti-FASN Polyclonal Antibody Pair

Supplier: Abnova
H00002194-PW3
ABNOH00002194-PW3EA 835 EUR
ABNOH00002194-PW3
Anti-FASN Polyclonal Antibody Pair
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-FASN (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-FASN (50 µg)

Type:
Antigen: FASN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR