Anti-SAP97 Rabbit Polyclonal Antibody

Supplier: BosterBio
PB9552
BSBTPB9552EA 550 EUR
BSBTPB9552
Anti-SAP97 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Disks large homolog 1(DLG1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: SAP97
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR