Anti-SPARC Rabbit Polyclonal Antibody

Supplier: BosterBio
PB9559
BSBTPB9559EA 579 EUR
BSBTPB9559
Anti-SPARC Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for SPARC(SPARC) detection. Tested with WB in Human.

Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR