Anti-FMO3 Antibody

Supplier: BosterBio
PB9590
BSBTPB9590EA 579 EUR
BSBTPB9590
Anti-FMO3 Antibody
Antibodies
Rabbit IgG polyclonal antibody for Dimethylaniline monooxygenase [N-oxide-forming] 3(FMO3) detection. Tested with WB in Human;Mouse; Rat.

Type: Primary
Antigen: FMO3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR