Anti-ACADSB Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3157-100EA 276 EUR
BIOV3157-100
Anti-ACADSB Rabbit Polyclonal Antibody
Antibodies
Short/branched chain acyl-CoA dehydrogenase(ACADSB) belongs to the acyl-CoA dehydrogenase family of enzymes catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. ACADSB catalyzes the degradation of L-isoleucine and has the greatest affinity towards (S)-2-methylbutyryl-CoA, isobutyryl-CoA and 2-methylhexanoyl-CoA as substrates.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACADSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR