Anti-ACADL Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3139-100EA 305 EUR
BIOV3139-100
Anti-ACADL Rabbit Polyclonal Antibody
Antibodies
ACADL belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes. This protein is involved in fatty acid and branched chain amino acid metabolism. ACADL catalyzes the initial step of mitochondria beta-oxidation of straight-chain fatty acid.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACADL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR