Anti-Smad5 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3465-100EA 299 EUR
BIOV3465-100
Anti-Smad5 Rabbit Polyclonal Antibody
Antibodies
Smad proteins, the mammalian homologs of the Drosophila Mothers against dpp (Mad), have been implicated as downstream effectors of TGFβ/BMP signaling. Smad1, Smad5, and Smad8 are effectors of BMP2 and BMP4 function while Smad2 and Smad3 are involved in TGF-β and activin-mediated growth modulation. Smad4 has been shown to mediate all of the above activities throμgh interaction with various Smad family members. Smad6 and Smad7 regulate the response to activin/TGFβ signaling by interfering with TGFβ-mediated phosphorylation of other Smad family members.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Smad5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR