Anti-Cystatin B Mouse Monoclonal Antibody [clone: RJMW2E7]

Supplier: BioVision
BIOV3487-100EA 333 EUR
BIOV3487-100
Anti-Cystatin B Mouse Monoclonal Antibody [clone: RJMW2E7]
Cystatin B (stefin B, NCPI) is a natural inhibitor of lysosomal cysteine proteases (cathepsins;Its expression seems to be connected to the regulation of proteolysis which is essential for many physiological and pathological processes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Cystatin B
Clonality: Monoclonal
Clone: RJMW2E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR