Anti-MMP-12 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3732-100EA 293 EUR
BIOV3732-100
Anti-MMP-12 Rabbit Polyclonal Antibody
Antibodies
The matrix metalloproteinases (MMP) are a family of peptidase enzymes responsible for the degradation of extracellular matrix components, including collagen, gelatin, fibronectin, laminin and proteoglycan. Transcription of MMP genes is differentially activated by phorbol ester, lipopolysaccharide (LPS) or staphylococcal enterotoxin B (SEB;MMP catalysis requires both calcium and zinc. MMP-12 may contribute to elastin degradation occurring in granulomatous skin diseases and may also participate in macrophage migration throμgh the epidermal and vascular basement membranes in inflammatory disorders.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MMP-12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR