Anti-MGMT Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3820-100EA 276 EUR
BIOV3820-100
Anti-MGMT Rabbit Polyclonal Antibody
MGMT (O-6-methylguanine-DNA methyltransferase) is a DNA repair protein that response to DNA damage by removing methly or alkyl groups from the O6 position of guanine. MGMT protects normal cells from cytotoxic effects and is important in drμg resistance to alkylating agents.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MGMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR