Anti-FKBP52/Hsp56 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3880-100EA 276 EUR
BIOV3880-100
Anti-FKBP52/Hsp56 Rabbit Polyclonal Antibody
FKBP52 (also known as FKBP4, Hsp56, FK506 binding protein 4, p52, FKBP59) is a 55kDa peptidyl-propyl isomerase. FKBP52 catalyzes the transition between cis- and trans- proline residues which is important in immunoregulation and proper protein folding and trafficking. FKBP52 may involve in the intracellular trafficking of hetero-oligomeric forms of the steroid hormone receptors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FKBP52/Hsp56
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR