Anti-DKK4 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3894-100EA 276 EUR
BIOV3894-100
Anti-DKK4 Rabbit Polyclonal Antibody
Xenopus Dickkopf (Dkk) was initially discovered as a Wnt antagonist that plays an important role in head formation. By far, four members of Dkk have been identified in mammals. Each Dkk molecule contains two conserved cysteine-rich domains which are separated by a 50-55 amino acid linker region. Dkk4 is a secreted glycoprotein and has three immune-reactive sites at 15-17kDa, 30-32kDa and at 40kDa.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DKK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR