Anti-CGI-58 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3901-100EA 276 EUR
BIOV3901-100
Anti-CGI-58 Rabbit Polyclonal Antibody
Antibodies
Comparative Gene Identification-58 (CGI-58) is a causal gene of Dorfman-Chanarin syndrome, a neutral lipid storage disease characterized by the presence of intracellular lipid droplets in tissues. CGI-58 comes from a large family of proteins characterized by an α/β hydrolase fold that activates adipose triglyceride lipase thus, may involve in lipid metabolism.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CGI-58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR