Anti-ALK-7 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3938-100EA 293 EUR
BIOV3938-100
Anti-ALK-7 Rabbit Polyclonal Antibody
Antibodies
ALK7 or ACVR1C is a type I receptor for TGFB superfamily of signaling protein. Upon ligand binding, type I receptors phosphorylate cytoplasmic SMAD transcription factors, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ALK-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR