Anti-S-Tag Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3990-100EA 276 EUR
BIOV3990-100
Anti-S-Tag Rabbit Polyclonal Antibody
S-tag is an epitope tag composed of a 15 residue peptide derived from pancreatic ribonuclease A

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: S-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S-Tag And S-Tag Fusion Proteins
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR