Anti-IGF-II Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5122-100EA 316 EUR
BIOV5122-100
Anti-IGF-II Rabbit Polyclonal Antibody
IGF-II (Insulin-like Growth Factor-II) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types. Human IGF-II is a 7.5 kDa protein containing 67 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGF-II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR