Anti-IL-6 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5145-100EA 299 EUR
BIOV5145-100
Anti-IL-6 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant rat IL-6. The antibody was purified by affinity chromatography employing immobilized rat IL-6 matrix.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR