Anti-M-CSF Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5237-100EA 276 EUR
BIOV5237-100
Anti-M-CSF Rabbit Polyclonal Antibody
Antibodies
Macrophage Colony Stimulating Factor (M-CSF) is produced by various cell types, including fibroblasts, microphages, and endothelial cells. M-CSF is involved in both the development and function of monocytes/microphages.

Recommended Dilutions: 0

Type: Primary
Antigen: M-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR