Anti-OPG Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5312-100EA 276 EUR
BIOV5312-100
Anti-OPG Rabbit Polyclonal Antibody
Human OPG (Osteoprotegerin), also known as OCIF (Osteoclastogenesis inhibitory factor), is a member of the TNF receptor superfamily. It specifically acts on bone tissues and increases bone mineral density and bone volume associated with a decrease of active osteoclast number. Human OPG is a 20 kDa protein, comprising of 174 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: OPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR