Anti-Beta-Galactosidase Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3231-100EA 310 EUR
BIOV3231-100
Anti-Beta-Galactosidase Rabbit Polyclonal Antibody
The GLB1 gene encodes beta-galactosidase-1 (EC 3.2.1.23), a lysosomal hydrolase that cleaves the terminal beta-galactose from ganglioside substrates and other glycoconjugates (Yoshida et al., 1991 [PubMed 1907800];Beta-galactosidase also occurs in a complex with neuraminidase (NEU1; MIM 608272) and protective protein/cathepsin A (PPCA; MIM 256540), which is a component of certain cell surface receptors. This protein has no beta-galactosidase catalytic activity, but plays functional roles in the formation of extracellular elastic fibers (elastogenesis) and in the development of connective tissue. Seems to be identical to the elastin-binding protein (EBP), a major component of the non- integrin cell surface receptor expressed on fibroblasts, smooth muscle cells, chondroblasts, leukocytes, and certain cancer cell types. In elastin producing cells, associates with tropoelastin intracellularly and functions as a recycling molecular chaperone which facilitates the secretions of tropoelastin and its assembly into elastic fibers. Cleaves beta-linked terminal galactosyl residues from gangliosides, glycoproteins, and glycosaminoglycans.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: Beta-Galactosidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR