Anti-Profilin-1 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3306-100EA 276 EUR
BIOV3306-100
Anti-Profilin-1 Rabbit Polyclonal Antibody
Profilin is a protein which maintains and shapes cytoskeleton by binding to actin. Profilin 1 has function in breast cancer cell motility, radial migration and glial cell adhesion.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Profilin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR