Anti-LXR Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV3641-100EA 299 EUR
BIOV3641-100
Anti-LXR Rabbit Polyclonal Antibody
Antibodies
Liver X receptors (LXRs) are oxysterol activated nuclear receptors which are involved in the regulation of genesis and metabolism of cholesterols and bile acids. LXRs form heterodimeric complexes with the retinoic acid receptors (RARs) once activated by their respective oxysterol ligands. Two known isoforms, alpha and beta, are differentially expressed; the alpha isoform predominantly expressed in liver, whereas beta isoform expression has been shown to be ubiquitous.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LXR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR